ADV371425119 AChemBlock

Methyl 1-Boc-3-azetidineacetate

  • MDL number MFCD11559001
  • Cas Number : 497160-14-2
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid/light yellow liqiuid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-boc-3-azetidineacetate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2238
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart