ADV371425122 AChemBlock

1-Methyl-3-azetidinecarboxylic acid

  • MDL number MFCD11520621
  • Cas Number : 875629-26-8
  • Molecular Weight : 115.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-3-azetidinecarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2243
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart