ADV371425124 AChemBlock

3-Aminomethyl-3-methyl-oxetane

  • MDL number MFCD08703640
  • Cas Number : 153209-97-3
  • Molecular Weight : 101.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid/light yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-aminomethyl-3-methyl-oxetane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2245
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart