ADV371425155 AChemBlock

trans-3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid

  • MDL number MFCD06657681
  • Cas Number : 939400-34-7
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-3-tert-butoxycarbonylamino-cyclobutanecarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2280
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart