ADV371425162 AChemBlock

3,6-Dichloro-4-(pyridin-4-yl)pyridazine

  • MDL number MFCD11040472
  • Cas Number : 202931-70-2
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid/yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-6-dichloro-4-pyridin-4-yl-pyridazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2288
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart