ADV371425175 AChemBlock

1-Boc-3-(iodomethyl)azetidine

  • MDL number MFCD09951816
  • Cas Number : 253176-94-2
  • Molecular Weight : 297.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellowish liquid/white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-iodomethyl-azetidine.html

Safety Information

signalword Danger
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2307
pictograms GHS05; GHS06; GHS07
hstatements H301;H312;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart