ADV371425177 AChemBlock

Methyl 3,3-difluoro-cyclobutanecarboxylate

  • MDL number MFCD17015922
  • Cas Number : 1234616-13-7
  • Molecular Weight : 150.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid/light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-3-difluoro-cyclobutanecarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2310
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart