ADV371425184 AChemBlock

5-Nitro-7-azaindole

  • MDL number MFCD06659682
  • Cas Number : 101083-92-5
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-nitro-7-azaindole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2317
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart