ADV371425192 AChemBlock

7-Chloroimidazo[1,2-a]pyridine

  • MDL number MFCD08275126
  • Cas Number : 4532-25-6
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/light-brown solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-chloroimidazo-1-2-a-pyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2325
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart