ADV371425206 AChemBlock

1,3-Dihydro-2H-pyrrolo[3,2-b]pyridin-2-one

  • MDL number MFCD02179615
  • Cas Number : 32501-05-6
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-3-dihydro-2h-pyrrolo-3-2-b-pyridin-2-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2343
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart