ADV371425214 AChemBlock

2-(1H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-isoindole-1,3(2H)-dione

  • MDL number MFCD11045038
  • Cas Number : 741686-49-7
  • Molecular Weight : 264.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-1h-pyrrolo-2-3-d-pyrimidin-4-yl-1h-isoindole-1-3-2h-dione.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2351
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart