ADV371425227 AChemBlock

Imidazo[1,2-a]pyridin-5-amine

  • MDL number MFCD05863377
  • Cas Number : 66358-23-4
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Yellow crystal
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/imidazo-1-2-a-pyridin-5-amine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2372
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart