ADV371425255 AChemBlock

exo-3-Benzyl-3-azabicyclo[3.1.0]hexane-6-methanol

  • MDL number MFCD16140332
  • Cas Number : 134575-07-8
  • Molecular Weight : 203.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/exo-3-benzyl-3-azabicyclo-3-1-0-hexane-6-methanol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2404
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart