ADV371425257 AChemBlock

exo-6-(Boc-aminomethyl)-3-azabicyclo[3.1.0]hexane

  • MDL number MFCD14581078
  • Cas Number : 134575-12-5
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/exo-6-boc-aminomethyl-3-azabicyclo-3-1-0-hexane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2406
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart