ADV371425258 AChemBlock

exo-3-Cbz-3-azabicyclo[3.1.0]hexane-6-methanol

  • MDL number MFCD17214343
  • Cas Number : 134575-14-7
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/exo-3-cbz-3-azabicyclo-3-1-0-hexane-6-methanol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2407
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart