ADV371425284 AChemBlock

3-Amino-5-bromo-pyridine-2-carboxylic acid isopropyl ester

  • MDL number MFCD17168419
  • Cas Number : 1313738-67-8
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-5-bromo-pyridine-2-carboxylic-acid-isopropyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2437
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart