ADV371425285 AChemBlock

5-Aminoisoindolin-1-one

  • MDL number MFCD10000828
  • Cas Number : 222036-66-0
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form gray solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-aminoisoindolin-1-one.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2439
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart