ADV371425306 AChemBlock

exo-6-Amino-3-Boc-3-azabicyclo[3.1.0]hexane

  • MDL number MFCD09832898
  • Cas Number : 273206-92-1
  • Molecular Weight : 198.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/exo-6-amino-3-boc-3-azabicyclo-3-1-0-hexane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2466
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart