ADV371425307 AChemBlock

6-Hydroxy-4-methyl-2-pyridone

  • MDL number MFCD00040464
  • Cas Number : 4664-16-8
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form cream powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-hydroxy-4-methyl-2-pyridone.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2468
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart