ADV371425319 AChemBlock

4-Chloro-5H-pyrrolo[3,2-d]pyrimidine

  • MDL number MFCD06658411
  • Cas Number : 84905-80-6
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form pale-yellow to brown solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-5h-pyrrolo-3-2-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2483
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart