ADV371425320 AChemBlock

4-Chloro-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD03030404
  • Cas Number : 5399-92-8
  • Molecular Weight : 154.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-1h-pyrazolo-3-4-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2484
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart