ADV371425325 AChemBlock

1H-Pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD09832891
  • Cas Number : 952182-02-4
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazolo-3-4-b-pyridine-5-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2489
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart