ADV371425330 AChemBlock

5-Boc-3-amino-4,6-dihydro-1H-pyrrolo[3,4-c]pyrazole

  • MDL number MFCD08275050
  • Cas Number : 398491-59-3
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-boc-3-amino-4-6-dihydro-1h-pyrrolo-3-4-c-pyrazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2494
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart