ADV371425332 AChemBlock

4-Chloro-5,7-dihydro-6H-pyrrolo[2,3-d]pyrimidin-6-one

  • MDL number MFCD09832903
  • Cas Number : 346599-63-1
  • Molecular Weight : 169.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid/white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-5-7-dihydro-6h-pyrrolo-2-3-d-pyrimidin-6-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2498
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart