ADV371425350 AChemBlock

endo-3-Amino-8-Boc-8-azabicyclo[3.2.1]octane

  • MDL number MFCD18428077
  • Cas Number : 207405-68-3
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/endo-3-amino-8-boc-8-azabicyclo-3-2-1-octane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3218
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart