ADV371425354 AChemBlock

5-Aminomethylpyrimidine

  • MDL number MFCD07776948
  • Cas Number : 25198-95-2
  • Molecular Weight : 109.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid/colorless solid/white crystalline solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-aminomethylpyrimidine.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3222
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart