ADV371425361 AChemBlock

4-Amino-2-hydroxypyridine

  • MDL number MFCD02682913
  • Cas Number : 38767-72-5
  • Molecular Weight : 110.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/yellowish solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-2-hydroxypyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3231
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart