ADV371425389 AChemBlock

1-Boc-hexahydro-1H-azepin-4-ol

  • MDL number MFCD07368610
  • Cas Number : 478832-21-2
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellowish solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-hexahydro-1h-azepin-4-ol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3271
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart