ADV371425390 AChemBlock

5-Cyanopyrimidine

  • MDL number MFCD00235052
  • Cas Number : 40805-79-6
  • Molecular Weight : 105.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-cyanopyrimidine.html

Safety Information

signalword Danger
pcodes P261;P264;P270;P271;P280;P301+312;P302+352;P304+340;P305+351+338;P310;P332+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3273
pictograms GHS05
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart