ADV371425392 AChemBlock

6-Chloro-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD10697157
  • Cas Number : 23002-51-9
  • Molecular Weight : 154.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid/light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-1h-pyrazolo-3-4-d-pyrimidine.html

Safety Information

signalword Warning
pcodes P261;P273;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3275
pictograms GHS07; GHS09
hstatements H315;H319;H335;H400
Availability details for
Quantity
  • Availability
  • Add to Cart