ADV371425397 AChemBlock

7-Azaindole-6-carboxylic acid

  • MDL number MFCD08272223
  • Cas Number : 898746-35-5
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-azaindole-6-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3280
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart