ADV371425398 AChemBlock

Methyl 2-pyrimidinecarboxylate

  • MDL number MFCD03426929
  • Cas Number : 34253-03-7
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-pyrimidinecarboxylate.html

Safety Information

signalword Danger
pcodes P260;P264;P280;P301+361+353;P304+340;P305+351+338;P310;P321;P363
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3283
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart