ADV371425404 AChemBlock

6-Chloroimidazo[1,2-b]pyridazine

  • MDL number MFCD07778345
  • Cas Number : 6775-78-6
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form gray solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloroimidazo-1-2-b-pyridazine.html

Safety Information

signalword Danger
pcodes P264;P270;P280;P301+312;P305+351+338;P310;P330
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3292
pictograms GHS05; GHS07
hstatements H302;H318
Availability details for
Quantity
  • Availability
  • Add to Cart