ADV371425409 AChemBlock

1-Boc-3-iodopyrrolidine

  • MDL number MFCD12031283
  • Cas Number : 774234-25-2
  • Molecular Weight : 297.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless or light-yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-iodopyrrolidine.html

Safety Information

signalword Warning
pcodes P261;P273;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3298
pictograms GHS07; GHS09
hstatements H302;H315;H319;H335;H400
Availability details for
Quantity
  • Availability
  • Add to Cart