ADV371425411 AChemBlock

5-Azaindole

  • MDL number MFCD00955936
  • Cas Number : 271-34-1
  • Molecular Weight : 118.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-azaindole.html

Safety Information

signalword Warning
pcodes P264;P280;P305+351+338;P337+313;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3300
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart