ADV371425413 AChemBlock

1-Methyl-4-(hydroxyethyl)piperidine

  • MDL number MFCD13188565
  • Cas Number : 21156-84-3
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-4-hydroxyethyl-piperidine.html

Safety Information

signalword Warning
pcodes P261;P264;P271;P280;P302+352;P304+340;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3302
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart