ADV371425417 AChemBlock

Methyl 5-aminopyrazine-2-carboxylate hydrochloride

  • MDL number MFCD18425624
  • Cas Number : 1417794-63-8
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-aminopyrazine-2-carboxylate-hydrochloride.html

Safety Information

signalword Danger
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3306
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart