ADV371425419 AChemBlock

3-Bromotetrahydrofuran

  • MDL number MFCD09056777
  • Cas Number : 19311-37-6
  • Molecular Weight : 151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid/light-yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-bromotetrahydrofuran.html

Safety Information

signalword Danger
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3309
pictograms GHS05; GHS07
hstatements H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart