ADV371425427 AChemBlock

2-(Methylthio)pyrimidin-4(3H)-one

  • MDL number MFCD00955672
  • Cas Number : 5751-20-2
  • Molecular Weight : 142.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white crystals
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-methylthio-pyrimidin-4-3h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3321
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart