ADV371425431 AChemBlock

6-Choro-5-azaindole

  • MDL number MFCD08690133
  • Cas Number : 74976-31-1
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-choro-5-azaindole.html

Safety Information

signalword Danger
pcodes P264;P270;P280;P301+310;P305+351+338;P310
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3327
pictograms GHS06; GHS05
hstatements H301;H318
Availability details for
Quantity
  • Availability
  • Add to Cart