ADV371425437 AChemBlock

2,6-Dichloro-9-isopropyl-8-methyl-9H-purine

  • MDL number MFCD18432767
  • Cas Number : 1313026-86-6
  • Molecular Weight : 245.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-6-dichloro-9-isopropyl-8-methyl-9h-purine.html

Safety Information

signalword Danger
pcodes P260;P264;P270;P273;P280;P301+312;P301+330+331;P303+361+353;P304+340;P305+351+338;P310;P363;P391
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3335
pictograms GHS05; GHS07; GHS09
hstatements H302;H314;H318;H401;H411
Availability details for
Quantity
  • Availability
  • Add to Cart