ADV371425438 AChemBlock

2-Methyl-3-nitropyridine

  • MDL number MFCD04114137
  • Cas Number : 18699-87-1
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-3-nitropyridine.html

Safety Information

signalword Danger
pcodes P264;P270;P273;P280;P301+312;P301+330+331;P303+361+353;P304+340;P305+351+338;P310;P363;P391
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3336
pictograms GHS05; GHS07; GHS09
hstatements H302;H314;H318;H401;H411
Availability details for
Quantity
  • Availability
  • Add to Cart