ADV371425439 AChemBlock

3-Methyl-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09607943
  • Cas Number : 116834-96-9
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-1h-pyrazolo-3-4-b-pyridine.html

Safety Information

signalword Warning
pcodes P264;P270;P280;P301+312;P305+351+338;P330;P337+313
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3338
pictograms GHS07
hstatements H302;H319
Availability details for
Quantity
  • Availability
  • Add to Cart