ADV371425440 AChemBlock

6-Bromoimidazo[1,2-b]pyridazine

  • MDL number MFCD12198219
  • Cas Number : 1159977-65-7
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form brown solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromoimidazo-1-2-b-pyridazine.html

Safety Information

signalword Warning
pcodes P273
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3339
pictograms GHS09
hstatements H400
Availability details for
Quantity
  • Availability
  • Add to Cart