ADV371425442 AChemBlock

5-Bromo-1,2-dihydro-1-methyl-2-oxo-nicotinic acid

  • MDL number MFCD17214399
  • Cas Number : 846048-15-5
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form grey solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-1-2-dihydro-1-methyl-2-oxo-nicotinic-acid.html

Safety Information

signalword Warning
pcodes P261;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P362;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3342
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart