ADV371425445 AChemBlock

Anacetrapib

  • MDL number MFCD16294903
  • Cas Number : 875446-37-0
  • Molecular Weight : 637.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form white crystalline powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/anacetrapib.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3348
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart