ADV371425447 AChemBlock

2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)ethan-1-ol

  • MDL number MFCD00100006
  • Cas Number : 103788-65-4
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-5-methyl-2-phenyl-1-3-oxazol-4-yl-ethan-1-ol.html

Safety Information

signalword Warning
pcodes P261;P264;P270;P271;P280;P301+312;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3350
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart