ADV371425448 AChemBlock

Methyl 5-azazindole-6-carboxylate

  • MDL number MFCD18425635
  • Cas Number : 1352394-18-3
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-azazindole-6-carboxylate.html

Safety Information

signalword Danger
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3352
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart