ADV371425451 AChemBlock

3-Bromo-6-methylpicolinonitrile

  • MDL number MFCD08059560
  • Cas Number : 717843-48-6
  • Molecular Weight : 197.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-6-methylpicolinonitrile.html

Safety Information

signalword Danger
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3357
pictograms GHS05
hstatements H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart