ADV371425457 AChemBlock

6-Isopropylpyridine-2-carboxylic acid

  • MDL number MFCD18256206
  • Cas Number : 337904-77-5
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-isopropylpyridine-2-carboxylic-acid.html

Safety Information

signalword Danger
pcodes P261;P264;P270;P271;P280;P301+312;P302+352;P304+340;P305+351+338;P310;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3365
pictograms GHS05; GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart